Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCA
Entrez Gene ID
827323
Gene Name
flowering time control protein FCA
Gene Aliases
AT4G16280, BETA, DELTA AND GAMMA, DL4180C, FCAALL.331, FLOWERING TIME CONTROL PROTEIN FCA ALPHA
Chromosome Location
Chr.4: 9206597 - 9214981 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341092.1 NP_001319956.1 9 - 10 1313 67
NM_117725.3 NP_193363.4 9 - 10 900 67
NM_148347.2 NP_680713.2 10 - 11 1109 67
NM_179211.3 NP_849542.2 8 - 9 3544 67

View Full Product Details