Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RXW8
Entrez Gene ID
842218
Gene Name
GDSL-like lipase/acylhydrolase superfamily protein
Gene Aliases
AT1G58520
Chromosome Location
Chr.1: 21729913 - 21738808 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333839.1 NP_001319269.1 11 - 12 2486 143
NM_001333840.1 NP_001319268.1 11 - 12 2525 143
NM_001333841.1 NP_001322290.1 11 - 12 2488 143
NM_001333842.1 NP_001322291.1 10 - 11 2595 143
NM_001333843.1 NP_001322289.1 11 - 12 2406 143
NM_148599.4 NP_683440.2 10 - 11 2212 143
GenBank mRNA KJ920364.1 - 10 - 11 2162 143

View Full Product Details