Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RXW8
Entrez Gene ID
842218
Gene Name
GDSL-like lipase/acylhydrolase superfamily protein
Gene Aliases
AT1G58520
Chromosome Location
Chr.1: 21729913 - 21738808 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001333839.1 NP_001319269.1 10 - 11 2305 136
NM_001333840.1 NP_001319268.1 10 - 11 2344 136
NM_001333841.1 NP_001322290.1 10 - 11 2307 136
NM_001333842.1 NP_001322291.1 9 - 10 2414 136
NM_001333843.1 NP_001322289.1 10 - 11 2225 136
NM_148599.4 NP_683440.2 9 - 10 2031 136
GenBank mRNA KJ920364.1 - 9 - 10 1981 136

View Full Product Details