Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NFD3
Entrez Gene ID
840071
Gene Name
Ribosomal L18p/L5e family protein
Gene Aliases
AT1G31817, F5M6.25, F5M6_25, NUCLEAR FUSION DEFECTIVE 3
Chromosome Location
Chr.1: 11414695 - 11416444 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102918.3 NP_564385.1 1 - 2 199 66
GenBank mRNA AY063845.1 - 1 - 2 140 66
AY117289.1 - 1 - 2 111 66

View Full Product Details