Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- XTH30
- Entrez Gene ID
-
840109
- Gene Name
- xyloglucan endotransglucosylase/hydrolase 30
- Gene Aliases
-
AT1G32170, F3C3.5, F3C3_5, XTR4, XYLOGLUCAN ENDOTRANSGLYCOSYLASE-RELATED PROTEIN, xyloglucan endotransglucosylase/hydrolase 30, xyloglucan endotransglycosylase 4
- Chromosome Location
-
Chr.1: 11575320 - 11578030 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_102950.4 |
NP_174496.1 |
3 - 4 |
608 |
117 |
GenBank mRNA |
AY062698.1 |
- |
3 - 4 |
607 |
117 |
View Full Product Details