Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSLD6
Entrez Gene ID
840110
Gene Name
cellulose synthase-like D6
Gene Aliases
AT1G32180, ATCSLD6, CELLULOSE SYNTHASE LIKE D6, F3C3.4, F3C3_4, cellulose synthase-like D6
Chromosome Location
Chr.1: 11586391 - 11590035 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332996.1 NP_001321681.1 3 - 4 1448 65
NM_102951.2 NP_174497.1 2 - 3 1195 65

View Full Product Details