Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TOM2A
Entrez Gene ID
840133
Gene Name
tobamovirus multiplication 2A
Gene Aliases
AT1G32400, F5D14.17, F5D14_17, tobamovirus multiplication 2A
Chromosome Location
Chr.1: 11689064 - 11691588 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036050.2 NP_001031127.1 5 - 6 1328 74
NM_102974.5 NP_564399.1 6 - 7 1042 74
NM_202223.3 NP_973952.1 6 - 7 1110 74
GenBank mRNA AB085684.1 - 6 - 7 896 74
AF334723.1 - 5 - 6 736 74
AF385687.1 - 6 - 7 952 74
AY088560.1 - 6 - 7 934 74

View Full Product Details