Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ESP3
Entrez Gene ID
840143
Gene Name
RNA helicase family protein
Gene Aliases
AT1G32490, EMB2733, EMBRYO DEFECTIVE 2733, ENHANCED SILENCING PHENOTYPE 3, F5D14.27, F5D14_27
Chromosome Location
Chr.1: 11742006 - 11749788 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198198.1 NP_001185127.1 25 - 26 3145 88
NM_102984.5 NP_174527.2 25 - 26 3170 88
GenBank mRNA AY074318.1 - 25 - 26 3093 88
AY133872.1 - 24 - 25 2947 88

View Full Product Details