Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FP6
Entrez Gene ID
830015
Gene Name
farnesylated protein 6
Gene Aliases
AT4G38580, ATFP6, F20M13.140, F20M13_140, HEAVY METAL ASSOCIATED ISOPRENYLATED PLANT PROTEIN 26, HIPP26, farnesylated protein 6
Chromosome Location
Chr.4: 18034131 - 18035922 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342507.1 NP_001320160.1 2 - 3 410 81
NM_120019.3 NP_195570.1 1 - 2 310 81
GenBank mRNA AF324677.2 - 1 - 2 222 81
AF326881.1 - 1 - 2 222 81
AF339701.1 - 1 - 2 77 81
AF380660.1 - 1 - 2 222 81
AY056095.1 - 1 - 2 77 81

View Full Product Details