Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GH9B16
Entrez Gene ID
830054
Gene Name
glycosyl hydrolase 9B16
Gene Aliases
AT4G38990, AtGH9B16, F19H22.90, F19H22_90, glycosyl hydrolase 9B16
Chromosome Location
Chr.4: 18168676 - 18170949 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342521.1 NP_001329141.1 1 - 2 409 70
NM_120059.3 NP_195610.3 1 - 2 409 70

View Full Product Details