Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC14
Entrez Gene ID
830073
Gene Name
Sec14p-like phosphatidylinositol transfer family protein
Gene Aliases
ARABIDOPSIS THALIANA SECRETION 14, AT4G39180, ATSEC14, SECRETION 14, T22F8.80, T22F8_80
Chromosome Location
Chr.4: 18243726 - 18248805 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001204034.2 NP_001190963.1 5 - 6 728 113
NM_001342526.1 NP_001329546.1 5 - 6 794 113
NM_120078.2 NP_195629.2 5 - 6 1089 113

View Full Product Details