Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- bZIP2
- Entrez Gene ID
-
816329
- Gene Name
- basic leucine-zipper 2
- Gene Aliases
-
AT2G18160, ATBZIP2, F8D23.6, F8D23_6, FLORAL TRANSITION AT THE MERISTEM3, FTM3, G-BOX BINDING FACTOR 5, GBF5, TRANSCRIPTION FACTOR GBF5, basic leucine-zipper 2
- Chromosome Location
-
Chr.2: 7898012 - 7899242 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_127373.2 |
NP_179408.1 |
1 - 1 |
1078 |
131 |
GenBank mRNA |
BT000700.1 |
- |
1 - 1 |
989 |
131 |
View Full Product Details