Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPC2
Entrez Gene ID
818860
Gene Name
phosphoenolpyruvate carboxylase 2
Gene Aliases
AT2G42600, ATPPC2, F14N22.13, F14N22_13, phosphoenolpyruvate carboxylase 2
Chromosome Location
Chr.2: 17734302 - 17739167 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001336987.1 NP_001324501.1 7 - 8 1617 65
NM_180041.5 NP_850372.4 8 - 9 1599 65
NM_180042.5 NP_850373.4 8 - 9 1307 65

View Full Product Details