Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GBF3
Entrez Gene ID
819235
Gene Name
G-box binding factor 3
Gene Aliases
AT2G46270, BZIP TRANSCRIPTION FACTOR, G-box binding factor 3, T3F17.8
Chromosome Location
Chr.2: 19000180 - 19003403 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337182.1 NP_001323457.1 8 - 9 873 83
NM_001337183.1 NP_001323456.1 8 - 9 873 83
NM_130190.3 NP_182150.1 8 - 9 888 83
NM_180118.2 NP_850449.1 8 - 9 888 83
GenBank mRNA AF370307.1 - 8 - 9 828 83
AY063093.1 - 7 - 8 595 83

View Full Product Details