Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIP-1
Entrez Gene ID
819236
Gene Name
TGF-beta receptor interacting protein 1
Gene Aliases
AT2G46280, T3F17.7, TGF-beta receptor interacting protein 1, TIF3I1, TRIP1
Chromosome Location
Chr.2: 19003158 - 19005732 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130191.6 NP_182151.1 2 - 3 545 62
NM_180119.3 NP_850450.1 3 - 4 655 62
NM_201968.1 NP_973697.1 2 - 3 514 62
GenBank mRNA AK226590.1 - 2 - 3 495 62

View Full Product Details