Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATSK12
Entrez Gene ID
819753
Gene Name
Protein kinase superfamily protein
Gene Aliases
ASKGAMMA, AT3G05840, F10A16.14, F10A16_14
Chromosome Location
Chr.3: 1739777 - 1743163 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111458.5 NP_187235.1 7 - 8 1294 64
NM_180189.4 NP_850520.1 7 - 8 1022 64
GenBank mRNA AK227034.1 - 7 - 8 1062 64
AK316673.1 - 7 - 8 1091 64
AY062713.1 - 7 - 8 1007 64
AY093347.1 - 6 - 7 816 64
X75431.1 - - 989 64

View Full Product Details