Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GGT1
Entrez Gene ID
830118
Gene Name
gamma-glutamyl transpeptidase 1
Gene Aliases
AT4G39640, T19P19.30, T19P19_30, gamma-glutamyl transpeptidase 1
Chromosome Location
Chr.4: 18399923 - 18403067 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120124.6 NP_195674.2 4 - 5 1261 105
NM_202988.4 NP_974717.1 3 - 4 1467 105
GenBank mRNA AY069872.1 - 4 - 5 1157 105
BT000495.1 - 3 - 4 1081 105

View Full Product Details