Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
iqd2
Entrez Gene ID
831696
Gene Name
IQ-domain 2
Gene Aliases
AT5G03040, F15A17.70, F15A17_70, IQ-domain 2
Chromosome Location
Chr.5: 709948 - 713133 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161221.1 NP_001154693.1 4 - 5 844 108
NM_001203282.1 NP_001190211.1 4 - 5 1057 108
NM_120382.4 NP_568110.2 4 - 5 1196 108
GenBank mRNA AY057527.1 - 4 - 5 815 108
AY143972.1 - 3 - 4 604 108

View Full Product Details