Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IMPA-9
Entrez Gene ID
831675
Gene Name
importin alpha isoform 9
Gene Aliases
AT5G03070, F15A17.100, F15A17_100, importin alpha isoform 9
Chromosome Location
Chr.5: 718136 - 721559 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120385.4 NP_195927.2 10 - 11 1559 59

View Full Product Details