Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LCV3
Entrez Gene ID
816360
Gene Name
like COV 3
Gene Aliases
AT2G18460, T30D6.3, T30D6_3, like COV 3
Chromosome Location
Chr.2: 8001234 - 8002998 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127402.4 NP_179436.3 5 - 6 769 121

View Full Product Details