Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MEE16
Entrez Gene ID
816380
Gene Name
RING/U-box superfamily protein
Gene Aliases
AT2G18650, MSF3.3, MSF3_3, maternal effect embryo arrest 16
Chromosome Location
Chr.2: 8086572 - 8088302 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127421.2 NP_179455.1 1 - 1 1306 87

View Full Product Details