Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTHFR1
Entrez Gene ID
825167
Gene Name
methylenetetrahydrofolate reductase 1
Gene Aliases
AT3G59970, methylenetetrahydrofolate reductase 1
Chromosome Location
Chr.3: 22151164 - 22154483 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115860.4 NP_191556.1 3 - 4 854 66
NM_180392.1 NP_850723.1 3 - 4 797 66
NM_180393.1 NP_850724.1 3 - 4 797 66
GenBank mRNA AF181966.1 - 3 - 4 777 66
AY070034.1 - 3 - 4 797 66
AY122922.1 - 3 - 4 715 66

View Full Product Details