Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDF1B
Entrez Gene ID
831318
Gene Name
peptide deformylase 1B
Gene Aliases
AT5G14660, ATDEF2, DEF2, peptide deformylase 1B
Chromosome Location
Chr.5: 4726641 - 4728746 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343352.1 NP_001332718.1 1 - 2 294 91
NM_121470.3 NP_196970.1 1 - 2 294 91
NM_180490.1 NP_850821.1 1 - 2 294 91
GenBank mRNA AY050879.1 - 1 - 2 294 91
AY096673.1 - 1 - 2 219 91

View Full Product Details