Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDF1B
Entrez Gene ID
831318
Gene Name
peptide deformylase 1B
Gene Aliases
AT5G14660, ATDEF2, DEF2, peptide deformylase 1B
Chromosome Location
Chr.5: 4726641 - 4728746 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343352.1 NP_001332718.1 2 - 3 430 119
NM_121470.3 NP_196970.1 2 - 3 430 119
NM_180490.1 NP_850821.1 2 - 3 430 119
GenBank mRNA AY050879.1 - 2 - 3 430 119
AY096673.1 - 2 - 3 355 119

View Full Product Details