Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PGLCT
Entrez Gene ID
831472
Gene Name
plastidic GLC translocator
Gene Aliases
AT5G16150, GLT1, GLUCOSE TRANSPORTER 1, T21H19.70, T21H19_70, plastidic GLC translocator
Chromosome Location
Chr.5: 5272476 - 5275815 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_121620.2 NP_568328.1 11 - 12 1352 104
NM_180497.2 NP_850828.1 11 - 12 1437 104
NM_203058.3 NP_974787.1 10 - 11 1524 104
GenBank mRNA AY058152.1 - 11 - 12 1352 104
AY091052.1 - 11 - 12 1336 104
AY117359.1 - 10 - 11 1250 104

View Full Product Details