Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ELF6
Entrez Gene ID
830303
Gene Name
Zinc finger (C2H2 type) family protein / transcription factor jumonji (jmj) family protein
Gene Aliases
AT5G04240, EARLY FLOWERING 6, F21E1.160, F21E1_160
Chromosome Location
Chr.5: 1169153 - 1174992 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120506.5 NP_196044.2 5 - 6 2259 126
GenBank mRNA AY664500.1 - 5 - 6 1868 126

View Full Product Details