Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDG38
Entrez Gene ID
830550
Gene Name
SET domain protein 38
Gene Aliases
AT5G06620, ATXR4, F15M7.15, F15M7_15, SET domain protein 38
Chromosome Location
Chr.5: 2033760 - 2036266 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342888.1 NP_001331184.1 4 - 5 908 91
NM_120745.3 NP_196280.2 5 - 6 811 91

View Full Product Details