Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERL2
Entrez Gene ID
830609
Gene Name
ERECTA-like 2
Gene Aliases
AT5G07180, ERECTA-like 2, T28J14.120, T28J14_120
Chromosome Location
Chr.5: 2227439 - 2233587 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342930.1 NP_001331523.1 14 - 15 1030 81
NM_120800.4 NP_196335.2 16 - 17 1585 81
GenBank mRNA AY244746.1 - 16 - 17 1415 81
FJ708772.1 - 16 - 17 1230 81

View Full Product Details