Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ERL2
Entrez Gene ID
830609
Gene Name
ERECTA-like 2
Gene Aliases
AT5G07180, ERECTA-like 2, T28J14.120, T28J14_120
Chromosome Location
Chr.5: 2227439 - 2233587 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342930.1 NP_001331523.1 20 - 21 1472 117
NM_120800.4 NP_196335.2 22 - 23 2027 117
GenBank mRNA AY244746.1 - 22 - 23 1857 117
FJ708772.1 - 22 - 23 1672 117

View Full Product Details