Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BON2
Entrez Gene ID
830621
Gene Name
Calcium-dependent phospholipid-binding Copine family protein
Gene Aliases
AT5G07300, BONZAI 2, T2I1.10, T2I1_10
Chromosome Location
Chr.5: 2299725 - 2303268 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_120812.4 NP_568180.1 9 - 10 1207 94
GenBank mRNA AF389301.1 - 9 - 10 971 94
BT010130.1 - 9 - 10 936 94

View Full Product Details