Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PMDH1
Entrez Gene ID
816808
Gene Name
peroxisomal NAD-malate dehydrogenase 1
Gene Aliases
AT2G22780, T30L20.4, T30L20_4, peroxisomal NAD-malate dehydrogenase 1
Chromosome Location
Chr.2: 9689295 - 9692154 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127843.5 NP_179863.1 4 - 5 914 93
GenBank mRNA AF428346.1 - 4 - 5 826 93
AK227067.1 - 4 - 5 824 93
BT003009.1 - 4 - 5 683 93

View Full Product Details