Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SQE2
Entrez Gene ID
816814
Gene Name
squalene epoxidase 2
Gene Aliases
AT2G22830, T20K9.4, T20K9_4, squalene epoxidase 2
Chromosome Location
Chr.2: 9723615 - 9726384 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127848.4 NP_179868.1 4 - 5 1494 98

View Full Product Details