Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HHP3
Entrez Gene ID
816949
Gene Name
heptahelical protein 3
Gene Aliases
AT2G24150, F27D4.6, F27D4_6, heptahelical protein 3
Chromosome Location
Chr.2: 10265271 - 10267607 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001335921.1 NP_001325271.1 1 - 2 479 81
NM_127976.5 NP_565564.1 2 - 3 594 81
GenBank mRNA AF370179.1 - 2 - 3 373 81
BT001975.1 - 2 - 3 332 81

View Full Product Details