Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PUP5
Entrez Gene ID
816956
Gene Name
purine permease 5
Gene Aliases
AT2G24220, ATPUP5, F27D4.13, F27D4_13, purine permease 5
Chromosome Location
Chr.2: 10300032 - 10301787 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_127983.3 NP_179999.2 2 - 2 1206 81

View Full Product Details