Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NPH4
Entrez Gene ID
832196
Gene Name
Transcriptional factor B3 family protein / auxin-responsive factor AUX/IAA-like protein
Gene Aliases
ARF7, AT5G20730, AUXIN RESPONSE FACTOR 7, AUXIN-REGULATED TRANSCRIPTIONAL ACTIVATOR 7, AUXIN-RESPONSIVE TRANSCRIPTIONAL ACTIVATOR 7, BIP, BIPOSTO, IAA21, IAA23, IAA25, MASSUGU 1, MSG1, NON-PHOTOTROPHIC HYPOCOTYL, T1M15.130, T1M15_130, TIR5, TRANSPORT INHIBITOR RESPONSE 5, indole-3-acetic acid inducible 21, indole-3-acetic acid inducible 23, indole-3-acetic acid inducible 25
Chromosome Location
Chr.5: 7016445 - 7022179 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122080.3 NP_568400.2 5 - 6 1079 63
NM_180715.1 NP_851046.1 5 - 6 1079 63
NM_180716.3 NP_851047.1 5 - 6 1145 63
GenBank mRNA AY669789.1 - 5 - 6 470 63

View Full Product Details