Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RHF2A
Entrez Gene ID
832260
Gene Name
RING-H2 group F2A
Gene Aliases
AT5G22000, RING-H2 FINGER PROTEIN RHF2A, RING-H2 group F2A
Chromosome Location
Chr.5: 7277182 - 7280344 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343714.1 NP_001331677.1 6 - 7 1149 101
NM_180719.4 NP_851050.1 8 - 9 1415 101
NM_180720.1 NP_851051.1 8 - 9 1275 101
GenBank mRNA AF079182.1 - 8 - 9 1326 101
AF324994.1 - 8 - 9 1292 101

View Full Product Details