The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085146.2 | NP_001078615.1 | 4 - 5 | 833 | 92 |
NM_001085147.2 | NP_001078616.1 | 4 - 5 | 334 | 92 | |
NM_001343793.1 | NP_001331558.1 | 3 - 4 | 947 | 92 | |
NM_122227.2 | NP_197712.2 | 4 - 5 | 334 | 92 | |
NM_180731.3 | NP_851062.2 | 4 - 5 | 854 | 92 | |
GenBank mRNA | AK228998.1 | - | 4 - 5 | 334 | 92 |