Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SCPL34
Entrez Gene ID
832385
Gene Name
serine carboxypeptidase-like 34
Gene Aliases
AT5G23210, MKD15.7, MKD15_7, serine carboxypeptidase-like 34
Chromosome Location
Chr.5: 7810645 - 7815089 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085146.2 NP_001078615.1 8 - 9 1467 53
NM_001085147.2 NP_001078616.1 8 - 9 968 53
NM_001343793.1 NP_001331558.1 7 - 8 1581 53
NM_122227.2 NP_197712.2 8 - 9 968 53
NM_180731.3 NP_851062.2 8 - 9 1488 53
GenBank mRNA AK228998.1 - 8 - 9 968 53

View Full Product Details