Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARAD2
Entrez Gene ID
834523
Gene Name
Exostosin family protein
Gene Aliases
ARABINAN DEFICIENT 2, ARABINOSYLTRANSFERASE, AT5G44930, K21C13.11, K21C13_11
Chromosome Location
Chr.5: 18138775 - 18142159 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_123861.3 NP_199306.1 2 - 3 1160 108
NM_180801.2 NP_851132.1 2 - 3 1160 108
GenBank mRNA AY048259.1 - 2 - 3 1148 108
BT002634.1 - 2 - 3 1145 108
KJ138648.1 - 2 - 3 1145 108

View Full Product Details