The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_124156.5 | NP_199593.2 | 4 - 5 | 1215 | 88 |
NM_180820.2 | NP_851151.1 | 4 - 5 | 1236 | 88 | |
GenBank mRNA | AY158083.1 | - | 4 - 5 | 886 | 88 |