Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EMF2
Entrez Gene ID
835198
Gene Name
VEFS-Box of polycomb protein
Gene Aliases
AT5G51230, AT5G51240, AtEMF2, CYR1, CYTOKININ RESISTANT 1, EMBRYONIC FLOWER 2, MWD22.18, MWD22_18, VEF2
Chromosome Location
Chr.5: 20823644 - 20829644 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125941.1 NP_001119413.1 10 - 11 1270 152
NM_124502.3 NP_199936.2 11 - 12 1262 152
NM_180837.3 NP_851168.1 11 - 12 1354 152
GenBank mRNA AB053171.1 - 11 - 12 1199 152
AY140086.1 - 11 - 12 1211 152
BT008388.1 - 10 - 11 951 152

View Full Product Details