Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EMF2
Entrez Gene ID
835198
Gene Name
VEFS-Box of polycomb protein
Gene Aliases
AT5G51230, AT5G51240, AtEMF2, CYR1, CYTOKININ RESISTANT 1, EMBRYONIC FLOWER 2, MWD22.18, MWD22_18, VEF2
Chromosome Location
Chr.5: 20823644 - 20829644 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125941.1 NP_001119413.1 5 - 6 839 85
NM_124502.3 NP_199936.2 6 - 7 831 85
NM_180837.3 NP_851168.1 6 - 7 923 85
GenBank mRNA AB053171.1 - 6 - 7 768 85
AY140086.1 - 6 - 7 780 85
BT008388.1 - 5 - 6 520 85

View Full Product Details