Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- HGO
- Entrez Gene ID
-
835494
- Gene Name
- homogentisate 1,2-dioxygenase
- Gene Aliases
-
2-DIOXYGENASE, 2-dioxygenase, AT5G54080, AtHGO, HOMOGENTISATE 1, MJP23.6, MJP23_6, homogentisate 1
- Chromosome Location
-
Chr.5: 21945865 - 21948913 on Build TAIR10.1
- Gene Symbol
- AT5G51970
- Entrez Gene ID
-
835272
- Gene Name
- GroES-like zinc-binding alcohol dehydrogenase family protein
- Gene Aliases
-
AT5G51970, MSG15.7, MSG15_7
- Chromosome Location
-
Chr.5: 21111445 - 21113403 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK230367.1 |
- |
- |
95 |
106 |
View Full Product Details