Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VIP4
Entrez Gene ID
836236
Gene Name
leo1-like family protein
Gene Aliases
AT5G61150, MAF19.15, MAF19_15, VERNALIZATION INDEPENDENCE 4
Chromosome Location
Chr.5: 24603634 - 24607725 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125508.2 NP_200923.2 4 - 5 975 100
NM_180906.2 NP_851237.1 4 - 5 975 100
GenBank mRNA BT002058.1 - 4 - 5 884 100
BT008386.1 - 4 - 5 778 100

View Full Product Details