Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATH13
Entrez Gene ID
836618
Gene Name
ABC2 homolog 13
Gene Aliases
A. THALIANA OXIDATIVE STRESS-RELATED ABC1-LIKE PROTEIN 1, ABC2 homolog 13, ARABIDOPSIS THALIANA ABC2 HOMOLOG 13, AT5G64940, ATATH13, ATOSA1, MXK3.17, MXK3_17, OSA1, OXIDATIVE STRESS-RELATED ABC1-LIKE PROTEIN 1
Chromosome Location
Chr.5: 25948954 - 25953823 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125893.5 NP_201299.2 11 - 12 1668 75
NM_180940.2 NP_851271.1 11 - 12 1668 75
GenBank mRNA AY059877.1 - 11 - 12 1651 75
AY064044.1 - 11 - 12 1627 75
AY096382.1 - 11 - 12 1506 75

View Full Product Details