Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATH13
Entrez Gene ID
836618
Gene Name
ABC2 homolog 13
Gene Aliases
A. THALIANA OXIDATIVE STRESS-RELATED ABC1-LIKE PROTEIN 1, ABC2 homolog 13, ARABIDOPSIS THALIANA ABC2 HOMOLOG 13, AT5G64940, ATATH13, ATOSA1, MXK3.17, MXK3_17, OSA1, OXIDATIVE STRESS-RELATED ABC1-LIKE PROTEIN 1
Chromosome Location
Chr.5: 25948954 - 25953823 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_125893.5 NP_201299.2 17 - 18 2276 69
NM_180940.2 NP_851271.1 17 - 18 2276 69
GenBank mRNA AY059877.1 - 17 - 18 2259 69
AY064044.1 - 17 - 18 2235 69
AY096382.1 - 17 - 18 2114 69

View Full Product Details