Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CP33
Entrez Gene ID
824403
Gene Name
chloroplast RNA-binding protein 33
Gene Aliases
AT3G52380, PDE322, PIGMENT DEFECTIVE 322, chloroplast RNA-binding protein 33
Chromosome Location
Chr.3: 19421397 - 19423097 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115098.3 NP_190806.1 3 - 4 1072 57
GenBank mRNA AY039607.1 - 3 - 4 877 57
AY078022.1 - 3 - 4 850 57
AY085279.1 - 3 - 4 1072 57

View Full Product Details