Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VSR1
Entrez Gene ID
824451
Gene Name
vacuolar sorting receptor homolog 1
Gene Aliases
ARABIDOPSIS THALIANA EPIDERMAL GROWTH FACTOR RECEPTOR-LIKE PROTEIN, AT3G52850, ATELP, ATELP1, ATVSR1, BP-80, BP80, BP80-1;1, BP80B, GFS1, Green fluorescent seed 1, VACUOLAR SORTING RECEPTOR 1;1, VSR-1, VSR1;1, binding protein of 80 kDa 1;1, vacuolar sorting receptor homolog 1
Chromosome Location
Chr.3: 19587756 - 19591947 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115145.2 NP_190853.1 1 - 2 526 75
GenBank mRNA U86700.1 - 1 - 2 445 75
Y07917.1 - - 455 75

View Full Product Details