Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPa4
Entrez Gene ID
824530
Gene Name
pyrophosphorylase 4
Gene Aliases
AT3G53620, AtPPa4, pyrophosphorylase 4
Chromosome Location
Chr.3: 19880504 - 19883743 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115222.3 NP_190930.1 3 - 4 445 76
GenBank mRNA AK226578.1 - 3 - 4 329 76

View Full Product Details