Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FBL17
Entrez Gene ID
824630
Gene Name
RNI-like superfamily protein
Gene Aliases
AT3G54650
Chromosome Location
Chr.3: 20225730 - 20229214 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115322.4 NP_567005.2 4 - 5 1390 153
GenBank mRNA AF462827.1 - 4 - 5 1084 153
AY142006.1 - 4 - 5 1058 153

View Full Product Details